1,1,1,3,3,3-Hexafluoroisopropyl methacrylate 99% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-Hexafluoroisopropylmethacrylate
B21447-06
B21447-14
CAAAB21447-03EA
44.92
CAD
CAAAB21447-03
CAAAB21447-06
CAAAB21447-14
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate 99% stabilised
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate
Purity: 99%
CAS No.: 3063-94-3
Molecular Formula: CH2=C(CH3)CO2CH(CF3)2
Formula Weight: 236.12
Flash Point: 14°(57°F)
Refractive Index: 1.3310
Boiling Point: 99°
UN No.: 1993
MDL No.: MFCD00040105
Formula:
C₇H₆F₆O₂ MW: 236.12 g/mol Boiling Pt: 99 °C Density: 1.302 Flash Pt: 14 °C (57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040105 CAS Number: 3063-94-3 EINECS: 221-309-9 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...