2,3,6-Trimethylquinoxaline 97%

Supplier: Thermo Scientific Chemicals
Warning

B21432-03 B21432-06
CAAAB21432-03EA 56.25 CAD
CAAAB21432-03 CAAAB21432-06
2,3,6-Trimethylquinoxaline 97%
2,3,6-Trimethylquinoxaline

Purity: 97% CAS No.: 17635-21-1 Molecular Formula: C11H12N2 Formula Weight: 172.23 Melting Point: 91-94° MDL No.: MFCD00047593

Formula: C₁₁H₁₂N₂
MW: 172.23 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00047593
CAS Number: 17635-21-1
EINECS: 241-618-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR