1,1-Diphenyl-2-propyn-1-ol 99%
Supplier: Thermo Scientific Chemicals
B21428-06
B21428-14
CAAAB21428-06EA
71.73
CAD
CAAAB21428-06
CAAAB21428-14
1,1-Diphenyl-2-propyn-1-ol 99%
1,1-Diphenyl-2-propyn-1-ol
Purity: 99%
CAS No.: 3923-52-2
Molecular Formula: C15H12O
Formula Weight: 208.26
Flash Point: 113°(235°F)
Melting Point: 44-48°
Boiling Point: 179-180°/18mm
Beilstein Reference: 514549
MDL No.: MFCD00041570
Formula:
C₁₅H₁₂O Boiling Pt: 179…180 °C/18 mm Melting Pt: 44…48 °C Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041570 CAS Number: 3923-52-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...