Ammonium iron(III) oxalate trihydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iron ammonium oxalate, Ammonium ferric oxalate, Ammonium iron oxalate trihydrate

B21411-22 B21411-36
CAAAAB21411-22EA 76.95 CAD
CAAAAB21411-22 CAAAAB21411-36
Ammonium iron(III) oxalate trihydrate 98%
Ammonium iron(III) oxalate trihydrate

Purity: 98% CAS No.: 13268-42-3 Molecular Formula: (NH4)3Fe(C2O4)3.3H2O Formula Weight: 428.07 (374.02anhy) UN No.: 3288 MDL No.: MFCD00150137

Formula: (NH₄)₃Fe(C₂O₄)₃·3H₂O
MW: 428.07 g/mol (374.02 anhy)
Density: 1.780
Storage Temperature: Ambient
MDL Number: MFCD00150137
CAS Number: 13268-42-3
EINECS: 238-090-0
UN: 2923
ADR: 8,III
Merck Index: 14,517

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR