Boron trifluoridedimethanol complex (50 - 52% BF₃) (w/w)
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21357-AK
B21357-AU
CAAAB21357-AKEA
71.26
CAD
CAAAB21357-AK
CAAAB21357-AU
Boron trifluoridedimethanol complex (50 - 52% BF₃) (w/w)
Boron trifluoridedimethanol complex
Fieser: 1,73 9,64
CAS No.: 2802-68-8
Molecular Formula: BF3.2CH3OH
Formula Weight: 131.89
Flash Point: 68°(154°F)
Boiling Point: 58-60°/4mm
UN No.: 2922
MDL No.: MFCD00071635
Formula:
BF₃.₂CH₃OH Boiling Pt: 58…60 °C/4 mm Density: 1.222 Flash Pt: 68 °C(154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00071635 CAS Number: 2802-68-8 EINECS: 220-543-9 UN: 2922 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...