Methyl-3-methylsalicylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 2-hydroxy-3-methylbenzoate
B21341-04
B21341-09
CAAAB21341-04EA
74.73
CAD
CAAAB21341-04
CAAAB21341-09
Methyl-3-methylsalicylate 97%
Methyl 3-methylsalicylate
Purity: 97%
CAS No.: 23287-26-5
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 28-30°
Boiling Point: 234-236°
MDL No.: MFCD00020051
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 234…236 °C Melting Pt: 28…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020051 CAS Number: 23287-26-5 EINECS: 245-557-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...