Iminostilbene 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
5H-Dibenzo[b,f]azepine
B21323-14
B21323-22
B21323-36
CAAAB21323-14EA
241.74
CAD
CAAAB21323-14
CAAAB21323-22
CAAAB21323-36
Iminostilbene 97%
Iminostilbene
Purity: 97%
CAS No.: 256-96-2
Molecular Formula: C14H11N
Formula Weight: 193.25
Melting Point: 197-199°
Beilstein Reference: 1343358
Formula:
C₁₄H₁₁N MW: 193.25 g/mol Melting Pt: 197…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005071 CAS Number: 256-96-2 EINECS: 205-970-0 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...