N-Ethylguanidinesulfate 98%

Supplier: Thermo Scientific Chemicals
Warning

B21315-06 B21315-14
CAAAB21315-06EA 61.78 CAD
CAAAB21315-06 CAAAB21315-14
N-Ethylguanidinesulfate 98%
N-Ethylguanidinesulfate

Purity: 98% CAS No.: 3482-86-8 Molecular Formula: [CH3CH2NHC(=NH)NH2]2.H2SO4 Formula Weight: 272.33 Odor: Odorless Melting Point: ca 245° dec. MDL No.: MFCD00013130

Formula: [CH₃CH₂NHC(=NH)NH₂]₂·H₂SO₄
Melting Pt: ca 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013130
CAS Number: 3482-86-8
EINECS: 222-467-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR