2-Cyano-4'-methylbiphenyl 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4'-Methyl-[1,1'-biphenyl]-2-carbonitrile

B21273-14 B21273-22 B21273-06
CAAAAB21273-14EA 243.32 CAD
CAAAAB21273-14 CAAAAB21273-22 CAAAB21273-06
2-Cyano-4'-methylbiphenyl 98+%
2-Cyano-4'-methylbiphenyl

Purity: 98+% CAS No.: 114772-53-1 Molecular Formula: C14H11N Formula Weight: 193.26 Melting Point: 49-53° Boiling Point: 320° UN No.: 3077 MDL No.: MFCD00151805

Formula: C₁₄H₁₁N
MW: 193.26 g/mol
Boiling Pt: >320 °C
Melting Pt: 49…53 °C
Density: 1.170
Storage Temperature: Ambient
MDL Number: MFCD00151805
CAS Number: 114772-53-1
EINECS: 422-310-9
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR