D-Tagatose 99%
Supplier: Thermo Scientific Chemicals
B21192-06
B21192-01
B21192-03
CAAAAB21192-06EA
782.1
CAD
CAAAAB21192-06
CAAAB21192-01
CAAAB21192-03
D-Tagatose 99%
D-Tagatose
Purity: 99%
CAS No.: 87-81-0
Molecular Formula: C6H12O6
Formula Weight: 180.16
Melting Point: 129-135°
Beilstein Reference: 1724555
Optical Rotation: -4.7° (c=1 in water)
MDL No.: MFCD00134449
Formula:
C₆H₁₂O₆ Melting Pt: 129…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134449 CAS Number: 87-81-0 EINECS: 201-772-3 Merck Index: 14,9030 |
Specification Test Results
Optical rotation | -4.7° (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...