4-MUG 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3S,4S,5R,6S)-3,4,5-Trihydroxy-6-((4-methyl-2-oxo-2H-chromen-7-yl)oxy)tetrahydro-2H-pyran-2-carboxylic acid, 4-methylumbelliferyl-beta-D-glucuronide, 4-Methylumbelliferyl-β-D-glucuronide

B21190-MD B21190-MF
CAAAB21190-MDEA 431.34 CAD
CAAAB21190-MD CAAAB21190-MF
4-MUG 98%
4-MUG

Purity: 98% CAS No.: 6160-80-1 Molecular Formula: C16H16O9 Formula Weight: 352.30 Storage Temperature: -30°C to -10°C Optical Rotation: -106° (c=0.1 in water) MDL No.: MFCD00036772

Formula: C₁₆H₁₆O₉
MW: 352.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00150472
CAS Number: 6160-80-1
EINECS: 228-186-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR