Ethyl dichlorophosphite 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21179-30
B21179-09
B21179-18
CAAAAB21179-30EA
1903.9
CAD
CAAAAB21179-30
CAAAB21179-09
CAAAB21179-18
Ethyl dichlorophosphite 98%
Ethyl dichlorophosphite
Purity: 98%
CAS No.: 1498-42-6
Molecular Formula: CH3CH2OPCl2
Formula Weight: 146.94
Storage Temperature: 2°C to 8°C
Flash Point: 4°(39°F)
Refractive Index: 1.4660
Odor: Acrid
Boiling Point: 117-118°
Beilstein Reference: 1697483
UN No.: 2924
MDL No.: MFCD00000525
Formula:
C₂H₅Cl₂OP Boiling Pt: 117…118 °C Density: 1.284 Flash Pt: 4 °C(39 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000525 CAS Number: 1498-42-6 EINECS: 216-098-5 UN: 2924 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...