2-Acetylbenzo[b]thiophene 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Acetylbenzothiophene

B21173-03 B21173-06
CAAAB21173-03EA 81.37 CAD
CAAAB21173-03 CAAAB21173-06
2-Acetylbenzo[b]thiophene 98%
2-Acetylbenzo[b]thiophene

Purity: 98% CAS No.: 22720-75-8 Molecular Formula: C10H8OS Formula Weight: 176.23 Melting Point: 86-88° MDL No.: MFCD00090217

Formula: C₁₀H₈OS
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00090217
CAS Number: 22720-75-8
EINECS: 245-177-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR