Diethyl (2-bromoethyl)phosphonate 97%

Supplier: Thermo Scientific Chemicals
Warning

B21156-06 B21156-14
CAAAAB21156-06EA 81.69 CAD
CAAAAB21156-06 CAAAAB21156-14
Diethyl (2-bromoethyl)phosphonate 97%
Diethyl (2-bromoethyl)phosphonate

Purity: 97% CAS No.: 5324-30-1 Molecular Formula: C6H14BrO3P Formula Weight: 245.06 Flash Point: 110°(230°F) Refractive Index: 1.4605 Boiling Point: 98-100°/1mm Beilstein Reference: 1766077 MDL No.: MFCD00000239

Formula: C₆H₁₄BrO₃P
MW: 245.06 g/mol
Boiling Pt: 98…100 °C/1 mm
Density: 1,275
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000239
CAS Number: 5324-30-1
EINECS: 226-193-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR