Ethyl-α-bromophenylacetate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21114-09
B21114-18
B21114-30
CAAAAB21114-09EA
120.87
CAD
CAAAAB21114-09
CAAAAB21114-18
CAAAAB21114-30
Ethyl-α-bromophenylacetate 97%
Ethyl α-bromophenylacetate
Purity: 97%
CAS No.: 2882-19-1
Molecular Formula: C10H11BrO2
Formula Weight: 243.10
Flash Point:
Formula:
C₁₀H₁₁BrO₂ Boiling Pt: 164 °C/30 mm Density: 1.389 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013536 CAS Number: 2882-19-1 EINECS: 220-735-2 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...