N-Fmoc-L-alanine monohydrate 98%

Supplier: Thermo Scientific Chemicals

B21107-06 B21107-14
CAAAB21107-06EA 72.68 CAD
CAAAB21107-06 CAAAB21107-14
N-Fmoc-L-alanine monohydrate 98%
N-Fmoc-L-alanine monohydrate

Purity: 98% CAS No.: 207291-76-7 Molecular Formula: C18H17NO4.H2O Formula Weight: 329.36 (311.34anhy) Storage Temperature: 2°C to 8°C Melting Point: ca 130° Beilstein Reference: 2225975 Optical Rotation: -19° (c=1 in DMF) MDL No.: MFCD00037139

Formula: C₁₈H₁₇NO₄·H₂O
Melting Pt: ca 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00037139
CAS Number: 207291-76-7
EINECS: 252-660-6

Order Now

Specification Test Results

Optical rotation -19° (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR