Diphenylphosphine oxide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21101-03
B21101-06
B21101-14
CAAAB21101-03EA
98.75
CAD
CAAAB21101-03
CAAAB21101-06
CAAAB21101-14
Diphenylphosphine oxide 97%
Diphenylphosphine oxide
Purity: 97%
CAS No.: 4559-70-0
Molecular Formula: C12H11OP
Formula Weight: 202.20
Melting Point: 51-56°
MDL No.: MFCD00002079
Formula:
C₁₂H₁₁OP Melting Pt: 51…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002079 CAS Number: 4559-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...