Diethyl (phthalimidomethyl)phosphonate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Diethyl ((1,3-dioxoisoindolin-2-yl)methyl)phosphonate, (Phthalimidomethyl)phosphonic Acid Diethyl Ester
B21096-06
B21096-14
CAAAB21096-06EA
88.01
CAD
CAAAB21096-06
CAAAB21096-14
Diethyl (phthalimidomethyl)phosphonate 97%
Diethyl (phthalimidomethyl)phosphonate
Purity: 97%
CAS No.: 33512-26-4
Molecular Formula: C13H16NO5P
Formula Weight: 297.25
Melting Point: 60-64°
MDL No.: MFCD00134313
Formula:
C₁₃H₁₆NO₅P MW: 297.25 g/mol Melting Pt: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134313 CAS Number: 33512-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...