N-Fmoc-L-serine ≥97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Ser-OH
B21079-03
B21079-06
CAAAB21079-03EA
61.94
CAD
CAAAB21079-03
CAAAB21079-06
N-Fmoc-L-serine ≥97%
N-Fmoc-L-serine
Purity: 97+%
CAS No.: 73724-45-5
Molecular Formula: C18H17NO5
Formula Weight: 327.34
Storage Temperature: 2°C to 8°C
Melting Point: 102-106°
Beilstein Reference: 4715791
Optical Rotation: +13.6° (c=1 in ethyl acetate)
MDL No.: MFCD00051928
Formula:
C₁₈H₁₇NO₅ MW: 327.34 g/mol Melting Pt: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051928 CAS Number: 73724-45-5 |
Specification Test Results
Optical rotation | +13.6° (c=1 in ethyl acetate) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...