4-Methylumbelliferyl-β-D-galactopyranoside 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methylumbelliferyl β-D-galactopyranoside, 4-Methyl-7-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one

B21075-01 B21075-03
CAAAB21075-01EA 85.16 CAD
CAAAB21075-01 CAAAB21075-03
4-Methylumbelliferyl-β-D-galactopyranoside 98%
4-Methylumbelliferyl-β-D-galactopyranoside

Purity: 98% CAS No.: 6160-78-7 Molecular Formula: C16H18O8 Formula Weight: 338.32 Storage Temperature: -30°C to -10°C Melting Point: ca 260° dec. Beilstein Reference: 94673 Optical Rotation: -64° (c=1 in pyridine, 24h) MDL No.: MFCD00036773

Formula: C₁₆H₁₈O₈
MW: 338.312 g/mol
Melting Pt: ∼ 260 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00036773
CAS Number: 6160-78-7
EINECS: 228-185-5

Order Now

Specification Test Results

Optical rotation -64° (c=1 in pyridine, 24h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR