N-Fmoc-glycine 98%
Supplier: Thermo Scientific Chemicals
B21050-03
B21050-06
CAAAB21050-03EA
43.77
CAD
CAAAB21050-03
CAAAB21050-06
N-Fmoc-glycine 98%
N-Fmoc-glycine
Purity: 98%
CAS No.: 29022-11-5
Molecular Formula: FmocNHCH2CO2H
Formula Weight: 297.32
Storage Temperature: 2°C to 8°C
Melting Point: 172-175°
Beilstein Reference: 2163967
MDL No.: MFCD00037140
Formula:
C₁₇H₁₅NO₄ Melting Pt: 172…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037140 CAS Number: 29022-11-5 EINECS: 249-373-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...