3-Methylbenzofuran-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
B21044-03
B21044-06
CAAAAB21044-14EA
815.99
CAD
CAAAAB21044-14
CAAAB21044-03
CAAAB21044-06
3-Methylbenzofuran-2-carboxylic acid 97%
3-Methylbenzofuran-2-carboxylic acid
Purity: 97%
CAS No.: 24673-56-1
Molecular Formula: C10H8O3
Formula Weight: 176.17
Melting Point: 192-197°
Beilstein Reference: 132104
MDL No.: MFCD00060511
Formula:
C₁₀H₈O₃ Melting Pt: 192…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060511 CAS Number: 24673-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...