N-Fmoc-L-leucine 98%
Supplier: Thermo Scientific Chemicals
B21040-06
B21040-14
CAAAB21040-06EA
70.94
CAD
CAAAB21040-06
CAAAB21040-14
N-Fmoc-L-leucine 98%
N-Fmoc-L-leucine
Purity: 98%
CAS No.: 35661-60-0
Molecular Formula: C21H23NO4
Formula Weight: 353.42
Storage Temperature: 2°C to 8°C
Melting Point: 152-155°
Beilstein Reference: 2178254
Optical Rotation: -25° (c=1 in DMF)
MDL No.: MFCD00037133
Formula:
C₂₁H₂₃NO₄ Melting Pt: 152…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037133 CAS Number: 35661-60-0 EINECS: 252-662-7 |
Specification Test Results
Optical rotation | -25° (c=1 in DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...