Biphenyl-4-sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Biphenyl-4-sulphonyl Chloride, Biphenyl-4-sulfonyl Chloride

B21031-06 B21031-14 B21031-22
CAAAB21031-06EA 93.54 CAD
CAAAB21031-06 CAAAB21031-14 CAAAB21031-22
Biphenyl-4-sulfonyl chloride 97%
Biphenyl-4-sulphonyl chloride

Purity: 97% CAS No.: 1623-93-4 Molecular Formula: C12H9ClO2S Formula Weight: 252.72 Melting Point: 112-117° Boiling Point: 200-205°/6mm UN No.: 3261 MDL No.: MFCD00078658

Formula: C₁₂H₉ClO₂S
Boiling Pt: 200…205 °C/6 mm
Melting Pt: 112…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00078658
CAS Number: 1623-93-4
EINECS: 216-609-1
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR