N-Fmoc-L-valine 98%

Supplier: Thermo Scientific Chemicals
Warning

B21030-06 B21030-14
CAAAB21030-06EA 66.68 CAD
CAAAB21030-06 CAAAB21030-14
N-Fmoc-L-valine 98%
N-Fmoc-L-valine

Purity: 98% CAS No.: 68858-20-8 Molecular Formula: C20H21NO4 Formula Weight: 339.39 Storage Temperature: 2°C to 8°C Melting Point: 142-147° Beilstein Reference: 2177443 Optical Rotation: -17° (c=1 in DMF) MDL No.: MFCD00037124

Formula: C₂₀H₂₁NO₄
MW: 339.4 g/mol
Melting Pt: 142…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00037124
CAS Number: 68858-20-8
EINECS: 272-515-0

Order Now

Specification Test Results

Optical rotation -17° (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR