Tetrakis(trimethylsilyl)silane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21020-03
B21020-06
CAAAB21020-03EA
70.31
CAD
CAAAB21020-03
CAAAB21020-06
Tetrakis(trimethylsilyl)silane 98%
Tetrakis(trimethylsilyl)silane
Purity: 98%
CAS No.: 4098-98-0
Molecular Formula: Si[Si(CH3)3]4
Formula Weight: 320.85
Melting Point: 266-268° subl.
MDL No.: MFCD00054859
Formula:
C₁₂H₃₆Si₅ Melting Pt: 266…268 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00054859 CAS Number: 4098-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...