4-Carboxyphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20954-03
B20954-06
B20954-14
CAAAB20954-03EA
59.41
CAD
CAAAB20954-03
CAAAB20954-06
CAAAB20954-14
4-Carboxyphenylboronic acid 97%
4-Carboxyphenylboronic acid
Purity: 97%
CAS No.: 14047-29-1
Molecular Formula: C7H7BO4
Formula Weight: 165.94
Melting Point: ca 220° dec.
Beilstein Reference: 3031088
Formula:
C₇H₇BO₄ Melting Pt: ca 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00151801 CAS Number: 14047-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...