4-Amino-3-chloro-5-nitrobenzotrifluoride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-6-nitro-4-(trifluoromethyl)aniline
B20938-03
B20938-06
CAAAB20938-03EA
30.49
CAD
CAAAB20938-03
CAAAB20938-06
4-Amino-3-chloro-5-nitrobenzotrifluoride 97%
4-Amino-3-chloro-5-nitrobenzotrifluoride
Purity: 97%
CAS No.: 57729-79-0
Molecular Formula: C7H4ClF3N2O2
Formula Weight: 240.58
Melting Point: 63-67°
UN No.: 2811
MDL No.: MFCD00042153
Formula:
C₇H₄ClF₃N₂O₂ MW: 240.58 g/mol Melting Pt: 63…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042153 CAS Number: 57729-79-0 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...