2-Methyl-1-phenyl-2-propen-1-ol ≥85%, tech.

Supplier: Thermo Scientific Chemicals

B20936-06 B20936-14
CAAAB20936-06EA 95.43 CAD
CAAAB20936-06 CAAAB20936-14 CAAAB20936-22
2-Methyl-1-phenyl-2-propen-1-ol ≥85%, tech.
2-Methyl-1-phenyl-2-propen-1-ol

Purity: 85% CAS No.: 4383-08-8 Molecular Formula: C10H12O Formula Weight: 148.21 MDL No.: MFCD00046630

Formula: C₁₀H₁₂O
MW: 148.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046630
CAS Number: 4383-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR