4-Hydroxy-4'-nitrostilbene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20913-03
B20913-06
CAAAB20913-03EA
87.85
CAD
CAAAB20913-03
CAAAB20913-06
4-Hydroxy-4'-nitrostilbene 98%
4-Hydroxy-4'-nitrostilbene
Purity: 98%
CAS No.: 19221-08-0
Molecular Formula: C14H11NO3
Formula Weight: 241.25
Melting Point: 205-209°
Beilstein Reference: 2505854
MDL No.: MFCD00017044
Formula:
C₁₄H₁₁NO₃ Melting Pt: 205…209 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017044 CAS Number: 19221-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...