1-Bromo-4-(trifluoromethylthio)benzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20874-03
B20874-06
CAAAB20874-03EA
58.46
CAD
CAAAB20874-03
CAAAB20874-06
1-Bromo-4-(trifluoromethylthio)benzene 97%
1-Bromo-4-(trifluoromethylthio)benzene
Purity: 97%
CAS No.: 333-47-1
Molecular Formula: C7H4BrF3S
Formula Weight: 257.07
Flash Point: 83°(181°F)
Refractive Index: 1.5170
Boiling Point: 199-200°
Beilstein Reference: 2361926
MDL No.: MFCD00040840
Formula:
C₇H₄BrF₃S Boiling Pt: 199…200 °C Density: 1.710 Flash Pt: 83 °C(181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040840 CAS Number: 333-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...