Methyl cyclopropanecarboxylate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20853-14
B20853-22
CAAAB20853-14EA
57.04
CAD
CAAAB20853-14
CAAAB20853-22
Methyl cyclopropanecarboxylate 98%
Methyl cyclopropanecarboxylate
Purity: 98%
CAS No.: 2868-37-3
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 22°(71°F)
Refractive Index: 1.4180
Boiling Point: 118-119°
Beilstein Reference: 2039388
UN No.: 3272
MDL No.: MFCD00001280
Formula:
C₅H₈O₂ Boiling Pt: 118…119 °C Density: 0.983 Flash Pt: 22 °C(71 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001280 CAS Number: 2868-37-3 EINECS: 220-690-9 UN: 1992 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...