4,4'-Diethoxyazobenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20839-03
B20839-06
CAAAB20839-03EA
46.45
CAD
CAAAB20839-03
CAAAB20839-06
4,4'-Diethoxyazobenzene 97%
4,4'-Diethoxyazobenzene
Purity: 97%
CAS No.: 588-52-3
Molecular Formula: C16H18N2O2
Formula Weight: 270.33
MDL No.: MFCD00053756
Formula:
C₁₆H₁₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00053756 CAS Number: 588-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...