D-(-)-Camphorsulfonylimine ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20823-03
B20823-06
B20823-14
CAAAB20823-03EA
68.89
CAD
CAAAB20823-03
CAAAB20823-06
CAAAB20823-14
D-(-)-Camphorsulfonylimine ≥98%
D-(-)-Camphorsulfonylimine
Purity: 98+%
CAS No.: 60886-80-8
Molecular Formula: C10H15NO2S
Formula Weight: 213.30
Melting Point: 226-228°
Beilstein Reference: 85296
MDL No.: MFCD00064576
Formula:
C₁₀H₁₅NO₂S Melting Pt: 226…228 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064576 CAS Number: 60886-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...