Ethylene glycol ethyl methyl ether 97% stabilised

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: 0.01% Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

B20819-09 B20819-18
CAAAB20819-09EA 79.63 CAD
CAAAB20819-09 CAAAB20819-18
Ethylene glycol ethyl methyl ether 97% stabilised
Ethylene glycol ethyl methyl ether

Stab. with 0.01% BHT


Purity: 97% CAS No.: 5137-45-1 Molecular Formula: C5H12O2 Formula Weight: 104.15 Flash Point: 2°(36°F) Refractive Index: 1.3870 Boiling Point: 100-102° UN No.: 3271 MDL No.: MFCD00048580

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 100…102 °C
Density: 0.850
Flash Pt: 2 °C (36 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048580
CAS Number: 5137-45-1
EINECS: 225-893-6
UN: 3271
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR