7-Nitro-1H-indole-2-carboxylic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: CRT0044876

B20786-03 B20786-06
CAAAB20786-03EA 167.48 CAD
CAAAB20786-03 CAAAB20786-06
7-Nitro-1H-indole-2-carboxylic acid 96%
7-Nitro-1H-indole-2-carboxylic acid

Purity: 96% CAS No.: 6960-45-8 Molecular Formula: C9H6N2O4 Formula Weight: 206.16 Melting Point: 264-266° MDL No.: MFCD00044720

Formula: C₉H₆N₂O₄
MW: 206.16 g/mol
Melting Pt: 264…266 °C
Storage Temperature: Ambient
MDL Number: MFCD00044720
CAS Number: 6960-45-8
EINECS: 230-154-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR