1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecane, 1,4-Cyclohexanedione bis(ethylene acetal), 1,4-Cyclohexanedione bisethylene ketal

B20784-09 B20784-30
CAAAB20784-09EA 70.31 CAD
CAAAB20784-09 CAAAB20784-18 CAAAB20784-30
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan 99%
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan

Purity: 99% CAS No.: 183-97-1 Molecular Formula: C10H16O4 Formula Weight: 200.23 Melting Point: 79-80° Boiling Point: 75-85°/0.5mm MDL No.: MFCD00010851

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Boiling Pt: 75…85 °C/0.5 mm
Melting Pt: 79…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00010851
CAS Number: 183-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR