4,4'-Diisopropylbiphenyl 97%
Supplier: Thermo Scientific Chemicals
B20779-03
B20779-06
CAAAB20779-03EA
52.61
CAD
CAAAB20779-03
CAAAB20779-06
4,4'-Diisopropylbiphenyl 97%
4,4'-Diisopropylbiphenyl
Purity: 97%
CAS No.: 18970-30-4
Molecular Formula: C18H22
Formula Weight: 238.37
Melting Point: 66°
MDL No.: MFCD00043533
Formula:
C₁₈H₂₂ Melting Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043533 CAS Number: 18970-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...