4-Allyl-1,6-heptadien-4-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Allylhepta-1,6-dien-4-ol

B20703-09 B20703-18 B20703-88
CAAAB20703-09EA 147.89 CAD
CAAAB20703-09 CAAAB20703-18 CAAAB20703-88
4-Allyl-1,6-heptadien-4-ol 99%
4-Allyl-1,6-heptadien-4-ol

Purity: 99% CAS No.: 10202-75-2 Molecular Formula: (CH2=CHCH2)3COH Formula Weight: 152.24 MDL No.: MFCD00039829

Formula: (CH₂=CHCH₂)₃COH
MW: 152.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039829
CAS Number: 10202-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR