Ethyl-3,5-dimethylisoxazole-4-carboxylate 97%

Supplier: Thermo Scientific Chemicals

B20642-09 B20642-88
CAAAB20642-09EA 210.14 CAD
CAAAB20642-09 CAAAB20642-88
Ethyl-3,5-dimethylisoxazole-4-carboxylate 97%
Ethyl-3,5-dimethylisoxazole-4-carboxylate

Purity: 97% CAS No.: 17147-42-1 Molecular Formula: C8H11NO3 Formula Weight: 169.18 Boiling Point: 87-89°/2mm MDL No.: MFCD00052557

Formula: C₈H₁₁NO₃
MW: 169.18 g/mol
Boiling Pt: 87…89 °C/2 mm
Storage Temperature: Ambient
MDL Number: MFCD00052557
CAS Number: 17147-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR