2,5-Dimethylpyrrole-3-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,5-Dimethyl-1H-pyrrole-3-carboxylic acid
B20635-03
B20635-06
B20635-14
CAAAB20635-03EA
169.06
CAD
CAAAB20635-03
CAAAB20635-06
CAAAB20635-14
2,5-Dimethylpyrrole-3-carboxylic acid 97%
2,5-Dimethylpyrrole-3-carboxylic acid
Purity: 97%
CAS No.: 57338-76-8
Molecular Formula: C7H9NO2
Formula Weight: 139.16
Melting Point: 199-201° dec.
MDL No.: MFCD00203874
Formula:
C₇H₉NO₂ MW: 139.16 g/mol Melting Pt: 199…201 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00203874 CAS Number: 57338-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...