Methyl-3-methyl-4-nitrobenzoate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 3-methyl-4-nitrobenzoate

B20627-06 B20627-14
CAAAB20627-06EA 71.42 CAD
CAAAB20627-06 CAAAB20627-14
Methyl-3-methyl-4-nitrobenzoate 97%
Methyl 3-methyl-4-nitrobenzoate

Purity: 97% CAS No.: 24078-21-5 Molecular Formula: C9H9NO4 Formula Weight: 195.18 Melting Point: 80-82° MDL No.: MFCD00085640

Formula: C₉H₉NO₄
MW: 195.18 g/mol
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00085640
CAS Number: 24078-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR