Ethyl-4-piperidinecarboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl piperidine-4-carboxylate, Ethyl isonipecotate, Ethyl 4-Piperidinecarboxylate

B20601-14 B20601-22
CAAAB20601-14EA 70.31 CAD
CAAAB20601-14 CAAAB20601-22
Ethyl-4-piperidinecarboxylate 98%
Ethyl 4-piperidinecarboxylate

Purity: 98% CAS No.: 1126-09-6 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Flash Point: 80°(176°F) Refractive Index: 1.4600 Boiling Point: 203-205° Beilstein Reference: 118419 MDL No.: MFCD00006003

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 203…205 °C
Density: 1.021
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006003
CAS Number: 1126-09-6
EINECS: 214-416-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR