Methyl-3-methoxythiophene-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

B20586-03 B20586-06
CAAAB20586-03EA 50.4 CAD
CAAAB20586-03 CAAAB20586-06 CAAAB20586-14
Methyl-3-methoxythiophene-2-carboxylate 97%
Methyl 3-methoxythiophene-2-carboxylate

Purity: 97% CAS No.: 62353-75-7 Molecular Formula: C7H8O3S Formula Weight: 172.20 Melting Point: 60-63° MDL No.: MFCD00102115

Formula: C₇H₈O₃S
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00102115
CAS Number: 62353-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR