3-Methyl-1,5-pentanediol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Methylpentane-1,5-diol

B20546-06 B20546-14
CAAAB20546-06EA 110.13 CAD
CAAAB20546-06 CAAAB20546-14
3-Methyl-1,5-pentanediol 99%
3-Methyl-1,5-pentanediol

Purity: 99% CAS No.: 4457-71-0 Molecular Formula: HOCH2CH2CH(CH3)CH2CH2OH Formula Weight: 118.18 Flash Point: 143°(289°F) Refractive Index: 1.454 Beilstein Reference: 1697331 MDL No.: MFCD00039547

Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Density: 0.97
Flash Pt: 143 °C(289 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039547
CAS Number: 4457-71-0
EINECS: 224-709-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR