2-Mercaptopyridine-N-oxide sodium salt, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20529-03
B20529-06
B20529-14
CAAAB20529-03EA
53.4
CAD
CAAAB20529-03
CAAAB20529-06
CAAAB20529-14
2-Mercaptopyridine-N-oxide sodium salt, anhydrous 98%
2-Mercaptopyridine N-oxide sodium salt
Purity: 98%
CAS No.: 3811-73-2
Molecular Formula: C5H4NNaOS
Formula Weight: 149.15
Melting Point: ca 290° dec.
Merck Reference: 14,7994
MDL No.: MFCD01941547
Formula:
C₅H₄NNaOS Melting Pt: ∼ 290 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01941547 CAS Number: 3811-73-2 EINECS: 223-296-5 Merck Index: 14,7994 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...