Hesperetin 97%
Supplier: Thermo Scientific Chemicals
B20528-03
B20528-06
B20528-14
CAAAB20528-03EA
53.56
CAD
CAAAB20528-03
CAAAB20528-06
CAAAB20528-14
Hesperetin 97%
Hesperetin
Purity: 97%
CAS No.: 520-33-2
Molecular Formula: C16H14O6
Formula Weight: 302.30
Melting Point: 227-232°
Beilstein Reference: 309850
Merck Reference: 14,4670
MDL No.: MFCD00075646
Formula:
C₁₆H₁₄O₆ Melting Pt: 227…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075646 CAS Number: 520-33-2 EINECS: 208-290-2 Merck Index: 14,4670 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...