Hyodeoxycholic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20506-06
B20506-14
B20506-22
CAAAB20506-06EA
113.44
CAD
CAAAB20506-06
CAAAB20506-14
CAAAB20506-22
Hyodeoxycholic acid 96%
Hyodeoxycholic acid
Purity: 96%
CAS No.: 83-49-8
Molecular Formula: C24H40O4
Formula Weight: 392.59
Melting Point: 197-199°
Optical Rotation: +8° (c=2 in ethanol)
MDL No.: MFCD00003681
Formula:
C₂₄H₄₀O₄ Melting Pt: 197…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003681 CAS Number: 83-49-8 EINECS: 201-483-2 Merck Index: 14,4857 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...