5-Chloro-2-thiophenesulfonamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20477-03
B20477-06
CAAAB20477-03EA
23.38
CAD
CAAAB20477-03
CAAAB20477-06
5-Chloro-2-thiophenesulfonamide 97%
5-Chloro-2-thiophenesulfonamide
Purity: 97%
CAS No.: 53595-66-7
Molecular Formula: C4H4ClNO2S2
Formula Weight: 197.67
Melting Point: 115-118°
MDL No.: MFCD00052584
Formula:
C₄H₄ClNO₂S₂ Melting Pt: 115…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052584 CAS Number: 53595-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...