1,3-Dibromo-3-methylbutane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20468-06
B20468-14
B20468-22
CAAAB20468-06EA
73.94
CAD
CAAAB20468-06
CAAAB20468-14
CAAAB20468-22
1,3-Dibromo-3-methylbutane 98%
1,3-Dibromo-3-methylbutane
Purity: 98%
CAS No.: 24443-15-0
Molecular Formula: (CH3)2CBrCH2CH2Br
Formula Weight: 229.95
Flash Point:
Formula:
C₅H₁₀Br₂ Density: 1.69 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060773 CAS Number: 24443-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...